missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SN-6
GREENER_CHOICE

Product Code. p-5546030
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18084302

Brand: Tocris Bioscience™ 2184/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective Na+/Ca2+ exchange inhibitor (reverse mode)

SN-6 is a selective Na+/Ca2+-exchange (NCX) inhibitor; displays some selectivity for NCX1. IC50 values are 2.9, 16 and 8.6 μM for inhibition of intracellular Na+-dependent 45Ca2+ uptake by cells expressing NCX1, NCX2 and NCX3 respectively. Has some affinity for mACh receptors (IC50 = 18 μM) but minimal activity against NCKX2 and various receptors and ion channels (IC50 > 30 μM). Preferentially blocks Ca2+ influx mode and is more selective for NCX isoforms than KB-R7943 (Cat. No. 1244). Anti-ischemic; potently protects against hypoxia-induced renal tubular cell damage (IC50 = 0.63 μM).
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type SN-6
Purity >99%
Molecular Weight (g/mol) 402.16
Inhibitors SN-6

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.