missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SN-6

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
SN-6 is a selective Na+/Ca2+-exchange (NCX) inhibitor; displays some selectivity for NCX1. IC50 values are 2.9, 16 and 8.6 μM for inhibition of intracellular Na+-dependent 45Ca2+ uptake by cells expressing NCX1, NCX2 and NCX3 respectively. Has some affinity for mACh receptors (IC50 = 18 μM) but minimal activity against NCKX2 and various receptors and ion channels (IC50 > 30 μM). Preferentially blocks Ca2+ influx mode and is more selective for NCX isoforms than KB-R7943 (Cat. No. 1244). Anti-ischemic; potently protects against hypoxia-induced renal tubular cell damage (IC50 = 0.63 μM).
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | SN-6 |
| Purity | >99% |
| Molecular Weight (g/mol) | 402.16 |
| Inhibitors | SN-6 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction