missing translation for 'onlineSavingsMsg'
Learn More

SN-38, MedChemExpress

Product Code. 16401977
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16401977 50 mg 50mg
16486286 100 mg 100mg
16461096 200 mg 200mg
16431696 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16401977 Supplier Medchem Express Supplier No. HY13704
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SN-38 (NK012) is an active metabolite of the Topoisomerase I inhibitor Irinotecan. SN-38 (NK012) inhibits DNA and RNA synthesis with IC50s of 0.077 and 1.3 μM, respectively.

SN-38 (NK012) is an active metabolite of the Topoisomerase I inhibitor Irinotecan. SN-38 (NK012) inhibits DNA and RNA synthesis with IC50s of 0.077 and 1.3 μM, respectively.

Specifications

Chemical Name or Material SN-38
CAS 86639-52-3
Color White
Purity Grade Notes Research
Molecular Formula C22H20N2O5
Quantity 50 mg
Synonym NK012
Solubility Information DMSO : 25 mg/mL (63.71 mM; Need ultrasonic)
SMILES O=C1[C@](O)(CC)C2=C(CO1)C(N3CC4=C(CC)C5=CC(O)=CC=C5N=C4C3=C2)=O
Molecular Weight (g/mol) 392.4
Formula Weight 392.4
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.