missing translation for 'onlineSavingsMsg'
Learn More

SN 2, Tocris Bioscience™

Product Code. p-4815656
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30002025

Brand: Tocris Bioscience 4907/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective TRPML3 channel activator

Selective TRPML3 channel activator (EC50 = 1.13 μM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 823218-99-1
Molecular Formula C17H21NO
Molecular Weight (g/mol) 255.361
InChI Key WKLZNTYMDOPBSE-UHFFFAOYSA-N
Synonym sn 2, sn-2, 5-mesityl-3-oxa-4-azatricyclo 5.2.1.0∼2,6∼ dec-4-ene, 5-mesityl-3-oxa-4-azatricyclo 5.2.1.02,6 dec-4-ene, d0g1ln, 5-mesityl-3-oxa-4-azatricyclo 5.2.1.0 2,6 dec-4-ene, 3a,4,5,6,7,7a-hexahydro-3-2,4,6-trimethylphenyl-4,7-methano-1,2-benzisoxazole
PubChem CID 11834987
SMILES CC1=CC(=C(C(=C1)C)C2=NOC3C2C4CCC3C4)C

Specifications

Color White
Quantity 10 mg
Formula Weight 255.35
Percent Purity >98%
Physical Form Solid
Chemical Name or Material SN 2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.