missing translation for 'onlineSavingsMsg'
Learn More

SN 2, MedChemExpress

Product Code. 16443167
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16443167

Brand: Medchem Express HY16696

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SN 2 is a potent activator of TRPML3 ion channel with an EC50 of 1.8 μM. SN 2 also acts as a potent inhibitor of Dengue virus 2 (DENV2) and Zika virus (ZIKV).

SN 2 is a potent activator of TRPML3 ion channel with an EC50 of 1.8 μM.

Specifications

Chemical Name or Material SN 2
Physical Form Solid
CAS 823218-99-1
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C17H21NO
Solubility Information DMSO : 100 mg/mL (391.62 mM; Need ultrasonic)
SMILES CC(C=C(C)C=C1C)=C1C2=NOC3C2C4CC3CC4
Molecular Weight (g/mol) 255.35
Formula Weight 255.35
Percent Purity 99.86%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.