Learn More
Motic™ SMZ161 Microscope
Product Code. p-8504006
Click to view available options
Head Style:
Binocular
Trinocular
Lighting Type:
None
Quartz Halogen with Intensity Control 12V/10w
Quartz Halogen with Intensity Control 12V/20w
Unit Size:
Each
Product Code. 15415422
Brand: Motic™ 1100201300052
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Convincing optical performance, a small footprint, and robust mechanics makes this the ideal stereo zoom microscope for teaching situations. Motic™ SMZ161 Microscope is constructed with light, ESD-material components.
  • Greenough optical system.
  • Sharp, three-dimensioned images and wide field of view.
  • Interpupillary distance: 50-75mm.
  • Ergonomically designed for long hours of viewing without discomfort.
  • Suitable for industrial applications, assembly and inspection of small parts or integrated circuits on PCB, LCD production line.
  • Coarse focusing system with tension adjustment.
Specifications
Includes Black and white stage plates and dust cover
Base Style Pole-type stand with transmitted illumination, 237x170mm
Magnification Power 0.75X to 4.5X
Objective Types Zoom system, ratio 1:6
Zoom Range 1:6
Product Type Stereo zoom microscope
Model SMZ161-BL
Head Style Binocular
Lighting Type Quartz Halogen with Intensity Control 12V/20w
Eyepiece WF10X/20 mm