missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMYD5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576863
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Retinoic acid (RA) represents the oxidized form of vitamin A and, via interactions with retinoic acid receptors (RARs), plays a crucial role in development, cellular growth and differentiation. The gene encoding RAI15 maps to human chromosome 2, which houses over 1,400 genes and comprises nearly 8% of the human genome. Harlequin icthyosis, a rare and morbid skin deformity, is associated with mutations in the ABCA12 gene, while the lipid metabolic disorder sitosterolemia is associated with defects in the ABCG5 and ABCG8 genes. Additionally, an extremely rare recessive genetic disorder, Alström syndrome, is caused by mutations in the ALMS1 gene, which maps to chromosome 2.
Specifications
| SMYD5 | |
| Polyclonal | |
| Unconjugated | |
| SMYD5 | |
| AW536703; NN8-4AG; Protein NN8 4AG; Protein NN8-4AG; Rai15; retinoic acid induced 15; retinoic acid responsive; retinoic acid responsive gene 1; retinoic acid-induced protein 15; RRG1; SET and MYND domain containing 5; SET and MYND domain-containing protein 5; SMYD family member 5; SMYD5; ZMYND23 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10322, 232187 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q3TYX3, Q6GMV2 | |
| SMYD5 | |
| Recombinant full length Human SMYD5. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction