Learn More
Invitrogen™ SMYD4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596631
Description
Immunogen sequence: MDLPVDEWKS YLLQKWASLP TSVQVTISTA ETLRDIFLHS SSLLQPEDEL FLKRLSKGYL VGKDSDAPLF YREEGNKKFQ EKDYTGAAVL YSKGVSHSRP NTEDMSLCHA NRSAALFHLG QYETCLKDIN RAQTHGYPER LQPKIMLRKA ECLVALGRLQ EASQTISDLE RNFTATPALA DVLPQTLQRN LHRLKMKMQE KDSLTESFPA ALAKTLEDAA LREENEQLSN ASSSIGLCVD PLKGRCLVAT KDILPGELLV; Positive Samples: Mouse kidney, Mouse brain, Mouse ovary, Rat kidney, Rat brain.
The specific function of the protein remains unknown.
Specifications
| SMYD4 | |
| Polyclonal | |
| Unconjugated | |
| SMYD4 | |
| G430029E23Rik; Kiaa1936; SET and MYND domain containing 4; SET and MYND domain-containing protein 4; Smyd4; ZMYND21 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 114826, 287525, 319822 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8BTK5, Q8IYR2 | |
| SMYD4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human SMYD4 (NP_4431602). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.