missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMYD3 Recombinant Rabbit Monoclonal Antibody (24GB2260)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557727
This item is not returnable.
View return policy
Description
SMYD3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
SMYD3 (histone-lysine N-methyltransferase) methylates Lys-4 of histone H3 and Lys-5 of histone H4. It induces di- and tri-methylation. SMYD3 plays an important role in transcriptional activation as a member of an RNA polymerase complex.
Specifications
| SMYD3 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SMYD3 | |
| 2410008A19Rik; bA74P14.1; bA74P14.1 (novel protein); histone-lysine N-methyltransferase SMYD3; KMT3E; SET and MYND domain containing 3; SET and MYND domain-containing protein 3; Smyd3; Zinc finger MYND domain-containing protein 1; zinc finger protein, subfamily 3A (MYND domain containing), 1; zinc finger, MYND domain containing 1; ZMYND1; ZNFN3A1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 498295, 64754, 69726 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2260 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9CWR2, Q9H7B4 | |
| SMYD3 | |
| A synthesized peptide derived from human SMYD3 (aa 251-314). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction