missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMYD3 Mouse anti-Human, Clone: 1B5C10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66330-1-IG-20UL
This item is not returnable.
View return policy
Description
SMYD3 (histone-lysine N-methyltransferase) methylates Lys-4 of histone H3 and Lys-5 of histone H4. It induces di- and tri-methylation. SMYD3 plays an important role in transcriptional activation as a member of an RNA polymerase complex.Specifications
| SMYD3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H7B4 | |
| SMYD3 | |
| SMYD3 Fusion Protein Ag2624 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1B5C10 | |
| Unconjugated | |
| SMYD3 | |
| SMYD3, ZMYND1, ZNFN3A1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 64754 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction