missing translation for 'onlineSavingsMsg'
Learn More

SMYD3 Mouse anti-Human, Clone: 1B5C10, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66330-1-IG-150UL

 View more versions of this product

Product Code. 16868105

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SMYD3 (histone-lysine N-methyltransferase) methylates Lys-4 of histone H3 and Lys-5 of histone H4. It induces di- and tri-methylation. SMYD3 plays an important role in transcriptional activation as a member of an RNA polymerase complex.
Specifications

Specifications

SMYD3
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H7B4
SMYD3
SMYD3 Fusion Protein Ag2624
150 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
1B5C10
Unconjugated
SMYD3
SMYD3, ZMYND1, ZNFN3A1
Mouse
Protein G
RUO
64754
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.