missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMYD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35194-100ul
This item is not returnable.
View return policy
Description
SMYD3 Polyclonal antibody specifically detects SMYD3 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| SMYD3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| bA74P14.1, bA74P14.1 (novel protein), EC 2.1.1, EC 2.1.1.43, FLJ21080, KMT3E, MGC104324, MYND domain containing 1, SET and MYND domain containing 3, SET and MYND domain-containing protein 3, Zinc finger MYND domain-containing protein 1, zinc finger protein, subfamily 3A (MYND domain containing), 1, ZMYND1, ZNFN3A1 | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human SMYD3 (NP_001161212.1).,, Sequence:, DRCLLGKEKLMRCSQCRVAKYCSAKCQKKAWPDHKRECKCLKSCKPRYPPDSVRLLGRVVFKLMDGAPSESEKLYSFYDLESNINKLTEDKKEGLRQLVMT | |
| 100 μL | |
| Cancer | |
| 64754 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction