missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMYD2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21290-1-AP-150UL
This item is not returnable.
View return policy
Description
SET domain-containing proteins, such as SMYD2, catalyze lysine methylation..Specifications
| SMYD2 | |
| Polyclonal | |
| Unconjugated | |
| SMYD2 | |
| HSKM B, KMT3C, Lysine N methyltransferase 3C, SMYD2, ZMYND14 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 226830, 56950 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8R5A0, Q9NRG4 | |
| SMYD2 | |
| SMYD2 Fusion Protein Ag15823 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction