missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SMYD2 Polyclonal specifically detects SMYD2 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | SMYD2 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 750 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | EC 2.1.1.43, Histone methyltransferase SMYD2, HSKM-Bzinc finger, MYND domain containing 14, KMT3CEC 2.1.1.-, Lysine N-methyltransferase 3C, MGC119305, SET and MYND domain containing 2, SET and MYND domain-containing protein 2, ZMYND14 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant SMYD2 (NP_064582.2; Met 1-His 433). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?