missing translation for 'onlineSavingsMsg'
Learn More

SMYD2 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18392614 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392614 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18392614 Supplier Novus Biologicals Supplier No. NBP299700AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SMYD2 Polyclonal specifically detects SMYD2 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMYD2
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias EC 2.1.1.43, Histone methyltransferase SMYD2, HSKM-Bzinc finger, MYND domain containing 14, KMT3CEC 2.1.1.-, Lysine N-methyltransferase 3C, MGC119305, SET and MYND domain containing 2, SET and MYND domain-containing protein 2, ZMYND14
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant SMYD2 (NP_064582.2; Met 1-His 433).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56950
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.