Learn More
Invitrogen™ SMYD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596108
Description
Immunogen sequence: MTIGRMENVE VFTAEGKGRG LKATKEFWAA DIIFAERAYS AVVFDSLVNF VCHTCFKRQE KLHRCGQCKF AHYCDRTCQK DAWLNHKNEC SAIKRYGKVP NENIRLAARI MWRVEREGTG LTEGCLVSVD DLQNHVEHFG EEEQKDLRVD VDTFLQYWPP QSQQFSMQYI SHIFGVINCN GFTLSDQRGL QAVGVGIFPN LGLVNHDCWP NCTVIFNNGN HEAVKSMFHT QMRIELRALG KISEGEELTV SYIDFLNVSE ERKRQLKKQY YFDCTCEHCQ KKLKDDLFLG VKDNPKPSQE VVKEMIQFSK DTLEKIDKAR SEGLYHEVVK LCRECLEKQE PVFADTNIYM; Positive Samples: Raji, MCF7, Mouse stomach, Rat heart, Rat stomach; Cellular Location: Cytoplasm, Nucleus.
Acts as a transcriptional repressor. Essential for cardiomyocyte differentiation and cardiac morphogenesis.
Specifications
| SMYD1 | |
| Polyclonal | |
| Unconjugated | |
| Smyd1 | |
| 4632404M21Rik; Bop; C78565; CD8 beta opposite; CD8beta opposite strand; CD8b-opposite; histone-lysine N-methyltransferase SMYD1; KMT3D; m-BOP; SET and MYND domain containing 1; SET and MYND domain-containing protein 1; SMYD1; Zinc finger protein BOP; zinc finger, MYND domain containing 18; Zmynd18; ZMYND22 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12180, 150572, 297333 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.74 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P97443, Q8NB12 | |
| Smyd1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-350 of human SMYD1 (NP_9380151). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.