missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMURF2 Monoclonal Antibody (1826CT865.51.28)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537753
This item is not returnable.
View return policy
Description
SMURF2 Monoclonal Antibody for Western Blot
SMURF2 is an E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. This protein interacts with SMAD1, SMAD2 and SMAD7 in order to trigger their ubiquitination and proteasome-dependent degradation. It enhances the inhibitory activity of SMAD7 and reduces the transcriptional activity of SMAD2. Coexpression of SMURF2 with SMAD1 results in considerable decrease in steady-state level of SMAD1 protein and a smaller decrease of SMAD2 level.
Specifications
| SMURF2 | |
| Monoclonal | |
| Unconjugated | |
| Smurf2 | |
| 2810411E22Rik; AI558114; AI649275; E3 ubiquitin ligase SMURF2; E3 ubiquitin-protein ligase SMURF2; HECT-type E3 ubiquitin transferase SMURF2; hSMURF2; RGD1310067; SMAD specific E3 ubiquitin protein ligase 2; SMAD ubiquitination regulatory factor 2; SMAD-specific E3 ubiquitin-protein ligase 2; SMURF2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 64750 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1826CT865.51.28 | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9HAU4 | |
| Smurf2 | |
| recombinant protein of human SMURF2. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction