missing translation for 'onlineSavingsMsg'
Learn More

SMURF2 Antibody - BSA Free, Novus Biologicals™

Product Code. 18397470 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397470 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18397470 Supplier Novus Biologicals Supplier No. NBP281916

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SMURF2 Polyclonal specifically detects SMURF2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMURF2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 2 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias DKFZp686F0270, E3 ubiquitin ligase SMURF2, E3 ubiquitin-protein ligase SMURF2, EC 6.3.2, EC 6.3.2.-, hSMURF2, MGC138150, SMAD specific E3 ubiquitin protein ligase 2, SMAD ubiquitination regulatory factor 2, SMAD-specific E3 ubiquitin-protein ligase 2
Gene Symbols SMURF2
Host Species Rabbit
Immunogen SMURF2 antibody was raised against a 17 amino acid peptide near the amino terminus of human SMURF2. The immunogen is located within amino acids 140 - 190 of SMURF2. Amino Acid Squence: SRLFDNDLPDGWEERR
Molecular Weight of Antigen 85 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64750
Test Specificity SMURF2 antibody is human, mouse and rat reactive. SMURF2 is predicted to not cross-react with SMURF1.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.