missing translation for 'onlineSavingsMsg'
Learn More

SMURF1 Antibody, Novus Biologicals™

Product Code. 18413079 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413079 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413079 Supplier Novus Biologicals Supplier No. H00057154D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SMURF1 Polyclonal antibody specifically detects SMURF1 in Human samples. It is validated for Western Blot, Proximity Ligation Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMURF1
Applications Western Blot, Proximity Ligation Assay
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Proximity Ligation Assay
Formulation PBS (pH 7.4)
Gene Accession No. NP_851994.1
Gene Alias E3 ubiquitin ligase SMURF1, EC 6.3.2, EC 6.3.2.-, hSMURF1, KIAA1625E3 ubiquitin-protein ligase SMURF1, SMAD specific E3 ubiquitin protein ligase 1, SMAD ubiquitination regulatory factor 1, Smad-specific E3 ubiquitin ligase 1, SMAD-specific E3 ubiquitin-protein ligase 1
Host Species Rabbit
Immunogen SMURF1 (NP_851994.1, 1 a.a. - 731 a.a.) full-length human protein. MSNPGTRRNGSSIKIRLTVLCAKNLAKKDFFRLPDPFAKIVVDGSGQCHSTDTVKNTLDPKWNQHYDLYVGKTDSITISVWNHKKIHKKQGAGFLGCVRLLSNAISRLKDTGYQRLDLCKLNPSDTDAVRGQIVVSLQTRDRIGTGGSVVDCRGLLENEGTVYEDSGPGRPLSCFMEEPAPYTDSTGAAAGGGNCRFVESPSQDQRLQAQRLRNPDVRGSLQTPQNRPHGHQSPELPEGYEQRTTVQGQVYFLHTQTGVSTWHDPRIPRDLNSVNCDELGPLPPGWEVRSTVSGRIYFVDHNNRTTQFTDPRLHHIMNHQCQLKEPSQPLPLPSEGSLEDEELPAQRYERDLVQKLKVLRHELSLQQPQAGHCRIEVSREEIFEESYRQIMKMRPKDLKKRLMVKFRGEEGLDYGGVAREWLYLLCHEMLNPYYGLFQYSTDNIYMLQINPDSSINPDHLSYFHFVGRIMGLAVFHGHYINGGFTVPFYKQLLGKPIQLSDLESVDPELHKSLVWILENDITPVLDHTFCVEHNAFGRILQHELKPNGRNVPVTEENKKEYVRLYVNWRFMRGIEAQFLALQKGFNELIPQHLLKPFDQKELELIIGGLDKIDLNDWKSNTRLKHCVADSNIVRWFWQAVETFDEERRARLLQFVTGSTRVPLQGFKALQGSTGAAGPRLFTIHLIDANTDNLPKAHTCFNRIDIPPYESYEKLYEKLLTAVEETCGFAVE
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 57154
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.