missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMUG1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | SMUG1 |
|---|---|
| Dilution | Western Blot 1:200-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18643491
|
Novus Biologicals
NBP2-93272-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18619820
|
Novus Biologicals
NBP2-93272-0.1ml |
0.1 mL |
£366.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMUG1 Polyclonal antibody specifically detects SMUG1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| SMUG1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Base Excision Repair, Cancer, DNA Repair | |
| PBS (pH 7.3), 50% glycerol | |
| 23583 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| EC 3.2.2, EC 3.2.2.-, FDG, HMUDG, MGC104370, single-strand selective monofunctional uracil DNA glycosylase, single-strand-selective monofunctional uracil-DNA glycosylase 1, UNG3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human SMUG1 (NP_055126.1). MPQAFLLGSIHEPAGALMEPQPCPGSLAESFLEEELRLNAELSQLQFSEPVGIIYNPVEYAWEPHRNYVTRYCQGPKEVLFLGMNPGPFGMAQTGVPFGEVSMVRDWLGIVGPVLTPPQEHPKRPVLGLECPQSEVSGARFWGFFRNLCGQPEVFFHHCFVHNLCPLLFLAPSGRNLTPAELPAKQREQLLGICDAALCRQVQLLGVRLVVGVGRLAEQRARRALAGLMPEVQVEGLLHPSPRNPQANKGWEAVAKERLNELGLLPLLLK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title