missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMS Recombinant Rabbit Monoclonal Antibody (23GB4640), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553781
This item is not returnable.
View return policy
Description
Catalyzes the production of spermine from spermidine and decarboxylated S-adenosylmethionine (dcSAM).
Specifications
| SMS | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SMS | |
| AI427066; Gy; gyro; MRSR; SMS; Spermidine aminopropyltransferase; spermine synthase; SpmST; SPMSY; SpS; SRS | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6611 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4640 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P52788 | |
| SMS | |
| A synthesized peptide derived from human Spermine synthase (100-150AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction