missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMS Recombinant Rabbit Monoclonal Antibody (1U7C7)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555560
This item is not returnable.
View return policy
Description
Immunogen Sequence: TYTNKNGSFA NLRIYPHGLV LLDLQSYDGD AQGKEEIDSI LNKVEERMKE LSQDSTGRVK RLPPIVRGGA IDRYWPTADG RLVEYDIDEV VYDEDSPYQN I.
Catalyzes the production of spermine from spermidine and decarboxylated S-adenosylmethionine (dcSAM).
Specifications
| SMS | |
| Recombinant Monoclonal | |
| 1.2 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P52788, P97355 | |
| SMS | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human Spermine synthase. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 1U7C7 | |
| Unconjugated | |
| SMS | |
| AI427066; Gy; gyro; MRSR; SMS; Spermidine aminopropyltransferase; spermine synthase; SpmST; SPMSY; SpS; SRS | |
| Rabbit | |
| Protein A | |
| RUO | |
| 20603, 363469, 6611 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction