missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMS Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583109
This item is not returnable.
View return policy
Description
Immunogen sequence: HSTLDFMLGA KADGETILKG LQSIFQEQGM AESVHTWQDH GYLATYTNKN GSFANLRIYP HGLVLLDLQS YDGDAQGKEE IDSILNKVEE RMKELS Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 90%.
Catalyzes the production of spermine from spermidine and decarboxylated S-adenosylmethionine (dcSAM).
Specifications
| SMS | |
| Polyclonal | |
| Unconjugated | |
| SMS | |
| AI427066; Gy; gyro; MRSR; SMS; Spermidine aminopropyltransferase; spermine synthase; SpmST; SPMSY; SpS; SRS | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6611 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P52788 | |
| SMS | |
| Recombinant protein corresponding to Human SMS. Recombinant protein control fragment (Product #RP-95184). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction