Learn More
SMS Mouse, anti-Human, CoraLite Plus 488, Clone: 1G8E9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-68040-100UL
Description
The SMS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SMS. This antibody recognizes human antigen. The SMS antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| SMS | |
| 1G8E9 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| MRSR, SMS, spermine synthase, SPMSY, SpS, SRS | |
| 100 μL | |
| 6611 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |