missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMR3A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560753
This item is not returnable.
View return policy
Description
Immunogen sequence: GPGRIQSHSL PPPYGPGYPQ PPSQPRPYPP GPPFFPVNSP TDPALPTPAP Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 46%.
SMR3A may play a role in protection or detoxification.
Specifications
| SMR3A | |
| Polyclonal | |
| Unconjugated | |
| SMR3A | |
| P-B1; PBI; PRL5; PROL5; proline rich 5 (salivary); proline- rich protein 5; proline-rich protein 5; Proline-rich protein PBI; protein homologous to salivary proline-rich protein P-B; PR-Vbeta1; SMR3A; submaxillary gland androgen regulated protein 3 homolog A; submaxillary gland androgen regulated protein 3A; submaxillary gland androgen-regulated protein 3A; variable coding sequence B1; Vcsb1; VCS-beta1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26952 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99954 | |
| SMR3A | |
| Recombinant protein corresponding to Human SMR3A. Recombinant protein control fragment (Product #RP-104369). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction