missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMPDL3B, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00027293-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MRLLAWLIFLANWGGARAEPGKFWHIADLHLDPDYKVSKDPFQVCPSAGSQPVPDAGPWGDYLCDSPWALINSSIYAMKEIEPEPDFILWTGDDTPHVPDEKLGEAAVLEIVERLTKLIREVFPDTKVYAALGNHDFHPKNQFPAGSNNIYNQIAELWKPWLSNESIALFKKGAFYCEKLPGPSGAGRIVVLNTNLYYTSNALTADMADPGQQFQWLEDVLTDASKAGDMVYIVGHVPPGFFEKTQNKAWFREGFNEKYLKVVRKHHRVIAGQFFGHHHTDSFRMLYDDAGVPISAMFITPGVTPWKTTLPGVVNGANNPAIRVFEYDRATLSLKVRSPAEARGGGWEGLKCITTFPHSQLIHLPLTTEPQEGSpecifications
| SMPDL3B | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_001009568.1 | |
| SMPDL3B | |
| SMPDL3B (NP_001009568.1, 1 a.a. ∼ 373 a.a) full-length human protein. | |
| RUO | |
| 27293 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| sphingomyelin phosphodiesterase, acid-like 3B | |
| ASML3B | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido