missing translation for 'onlineSavingsMsg'
Learn More

SMPDL3A Antibody, HRP, Novus Biologicals™

Product Code. 18355716 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18355716 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18355716 Supplier Novus Biologicals Supplier No. NBP300152H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SMPDL3A Polyclonal specifically detects SMPDL3A in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMPDL3A
Applications ELISA
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias 0610010C24Rik, acid sphingomyelinase-like phosphodiesterase 3a, ASM3A, ASML3a, ASM-like phosphodiesterase 3a, EC 3.1.4, EC 3.1.4.-, FLJ20177, sphingomyelin phosphodiesterase, acid-like 3A, yR36GH4.1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant SMPDL3A (Q92484; Met1-Tyr453).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 10924
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.