missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SMPD4 Polyclonal antibody specifically detects SMPD4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | SMPD4 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:200-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | KIAA1418, NET13, Neutral Sphingomyelinase 3, Neutral Sphingomyelinase III, NSMASE3, NSMASE-3, SKNY Protein, Sphingomyelin Phosphodiesterase 4, Sphingomyelin Phosphodiesterase 4, Neutral Membrane (Neutral Sphingomyelinase-3) |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 600-800 of human SMPD4 (NP_060421.2). ITQAKHTAKSISDQCAESPAGHSFLSWLGFSSMDTNGSYTANDLDEMGQDSVRKTDEYLEKALEYLRQIFRLSEAQLRQFTLALGTTQDENGKKQLPDCIVGEDGLILTPLGRYQIINGLRRFEIEYQGDPELQPIRSYEIASLVRTLFRLSSAINHRFAGQMAALCSRDDFLGSFCRYHLTEPGLASRHLLSPVGRRQVA |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?