missing translation for 'onlineSavingsMsg'
Learn More

SMPD3/NSMase2 Antibody [DyLight 405], Novus Biologicals™

Product Code. 30732834 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30732834 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732834 Supplier Novus Biologicals Supplier No. AF7184E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

SMPD3/NSMase2 Polyclonal antibody specifically detects SMPD3/NSMase2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMPD3/NSMase2
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias EC 3.1.4.12, FLJ22593, MGC138443, Neutral sphingomyelinase 2, Neutral sphingomyelinase II, nSMase2, nSMase-2, sphingomyelin phosphodiesterase 3, sphingomyelin phosphodiesterase 3, neutral membrane (neutral sphingomyelinaseII)
Host Species Sheep
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human SMPD3, Val2-Ala655, Accession # Q9NY59
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55512
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.