missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMPD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553732
This item is not returnable.
View return policy
Description
Immunogen sequence: QFIHHTSKKA DVVLLCGDLN MHPEDLGCCL LKEWTGLHDA YLETRDFKGS EEGNTMVPKN CYVSQQELKP FPFGVRIDYV LYKAVSGFYI SCKSFETTTG FDPHRGTPLS DHEALMATLF VRHSPPQQNP SST Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 82%.
Converts sphingomyelin to ceramide. Hydrolyze 1-acyl-2-lyso-sn-glycero-3-phosphocholine (lyso-PC) and 1-O-alkyl-2-lyso-sn-glycero-3-phosphocholine (lyso-platelet-activating factor). The physiological substrate seems to be Lyso-PAF.
Specifications
| SMPD2 | |
| Polyclonal | |
| Unconjugated | |
| Smpd2 | |
| AW108287; ISC1; Lyso-PAF-PLC; lyso-platelet-activating factor-phospholipase C; neutral sphingomyelinase; neutral sphingomyelinase 1; NSMA; NSMASE; N-SMase; nSMase1; SMPD2; Sphingomyelin phosphodiesterase 2; sphingomyelin phosphodiesterase 2, neutral; sphingomyelin phosphodiesterase 2, neutral membrane; sphingomyelin phosphodiesterase 2, neutral membrane (neutral sphingomyelinase) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6610 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O60906 | |
| Smpd2 | |
| Recombinant protein corresponding to Human SMPD2. Recombinant protein control fragment (Product #RP-91103). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction