missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMPD1 Antibody (OTI3H7), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74240G
This item is not returnable.
View return policy
Description
SMPD1 Monoclonal specifically detects SMPD1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| SMPD1 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SMPD1 | |
| Recombinant protein fragment corresponding to amino acids 62-341 of human SMPD1 (NP_000534) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI3H7 | |
| Western Blot, Immunohistochemistry | |
| Acid sphingomyelinase, ASMASE, ASMsphingomyelin phosphodiesterase, EC 3.1.4.12, NPD, sphingomyelin phosphodiesterase 1, acid lysosomal | |
| Mouse | |
| Affinity Purified | |
| Neuroscience | |
| 6609.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction