missing translation for 'onlineSavingsMsg'
Learn More

SMPD1 Antibody [DyLight 594], Novus Biologicals™

Product Code. 30736539 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30736539 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736539 Supplier Novus Biologicals Supplier No. AF5348M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

SMPD1 Polyclonal antibody specifically detects SMPD1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMPD1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Acid sphingomyelinase, ASMASE, ASMsphingomyelin phosphodiesterase, EC 3.1.4.12, NPD, sphingomyelin phosphodiesterase 1, acid lysosomal
Host Species Goat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human SMPD1, His62-Pro628, Accession # NP_000534
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6609
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.