missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMOX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561432
This item is not returnable.
View return policy
Description
Immunogen sequence: QGFTDVTVLE ASSHIGGRVQ SVKLGHATFE LGATWIHGSH GNPIYHLAEA NGLLEETTDG ERSVGRISLY SKNGVACYLT NHGRRIPKDV VEEFSDLYN Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 98%.
Polyamines are ubiquitous polycationic alkylamines which include spermine, spermidine, putrescine, and agmatine. These molecules participate in a broad range of cellular functions which include cell cycle modulation, scavenging reactive oxygen species, and the control of gene expression. These molecules also play important roles in neurotransmission through their regulation of cell-surface receptor activity, involvement in intracellular signalling pathways, and their putative roles as neurotransmitters. This gene encodes an FAD-containing enzyme that catalyzes the oxidation of spermine to spermadine and secondarily produces hydrogen peroxide. Multiple transcript variants encoding different isoenzymes have been identified for this gene, some of which have failed to demonstrate significant oxidase activity on natural polyamine substrates.
Specifications
| SMOX | |
| Polyclonal | |
| Unconjugated | |
| SMOX | |
| B130066H01Rik; C20orf16; flavin containing amine oxidase; flavin-containing spermine oxidase; PAO; PAO1; PAO-1; PAOH; PAOh1; Polyamine oxidase 1; putative cyclin G1 interacting protein; SMO; Smox; Spermine oxidase; UNQ3039/PRO9854 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54498 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NWM0 | |
| SMOX | |
| Recombinant protein corresponding to Human SMOX. Recombinant protein control fragment (Product #RP-99298). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction