missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Smoothelin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115934
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Smoothelin.
Smoothelin is a constituent of the smooth muscle cell (SMC) cytoskeleton. Antibodies directed to smoothelin are a useful tool to monitor SMC differentiation. Smoothelin is exclusively expressed in fully differentiated (contractile) SMCs. RNA and protein analyses reveal a broad species distribution of this protein. Cells with SMC-like characteristics, such as myofibroblasts, myoepithelial cells, and skeletal and cardiac muscle do not contain smoothelin. Confocal scanning laser microscopy of tissue sections, as well as transfection studies, show a filamentous organization of smoothelin that is different from that of desmin and vimentin. Smoothelin colocalizes with actin stress fibers. Two tissue specific isoforms have been identified: a 59 kDa isoform specific for visceral SMC; and a 100 kDa specific for vascular SMC. Human smoothelin is encoded by a single copy gene which is located on chromosome 22.
Specifications
| Smoothelin | |
| Polyclonal | |
| Unconjugated | |
| SMTN | |
| FLJ35365; FLJ38597; smoothelin; smoothelin-B; smsmo; SMTN | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 289734, 29856, 6525 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P53814, Q921U8 | |
| SMTN | |
| A synthesized peptide derived from human SMTN(Accession P53814), corresponding to amino acid residues S825-P875. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction