missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Smooth Muscle Differentiation Supplement (SMDS)

Product Code. 10276293
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10276293 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10276293 Supplier Gibco™ Supplier No. S0085

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Smooth Muscle Differentiation Supplement (SMDS)

A sterile, concentrated (100X) solution intended for use with Medium 231 to enhance differentiation of human smooth muscle cells. Each 5 ml bottle of SMDS contains the correct amount of differentiation supplement for a 500 ml bottle of Medium 231. When a 500 ml bottle of medium is supplemented with SMDS, the final concentrations of the components in the supplemented medium are:
  • fetal bovine serum, 1% v/v
  • heparin, 30μg/ml
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage minus 20 degrees C
    Cell Type Smooth Muscle Cells
    Concentration 100 X
    Form Liquid
    Product Type Differentiation Supplement
    Sterility Sterile
    For Use With (Application) Cell Differentiation
    Product Line Cascade Biologics
    Quantity 5 mL
    Shipping Condition Dry Ice
    Species Human
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.