missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ Smooth Muscle Growth Medium-2 BulletKit™
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Description
SmGM™-2 BulletKit™ is a complete kit optimized for the cultivation of primary derived cells from human smooth muscle cell systems, including prostate, umbilical, uterine, aortic/coronary, arterial, intestinal and bronchial/pulmonary cells. The kit contains 5% Fetal Bovine Serum.
Order Info
dimProductType
Specifications
Specifications
| Cell Type | Smooth Muscle Cells |
| Product Type | Growth Medium |
| For Use With (Application) | Maintenance and expansion of human smooth muscle cells |
| Quantity | 1 Kit |
| Content And Storage | 500mL bottle of Smooth Muscle Cell Basal Medium Growth supplements:
|
Safety and Handling
Product Identifier
- SmGM-2 Bullet Kit
- Danger
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : Gentamicin, sulfate(salt)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction