missing translation for 'onlineSavingsMsg'
Learn More

SMND-309, MedChemExpress

Product Code. p-200098648
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16498227

Brand: Medchem Express HY13056

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SMND-309 is a metabolite of salvianolic acid B, and exhibits neuroprotective effects in cultured neurons and in permanent middle cerebral artery occlusion rats.

SMND-309 is a metabolite of salvianolic acid B, and exhibits neuroprotective effects in cultured neurons and in permanent middle cerebral artery occlusion rats.

Specifications

Chemical Name or Material SMND-309
CAS 1065559-56-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H14O8
Quantity 5 mg
Solubility Information DMSO : ≥ 3.7 mg/mL (10.33 mM)
SMILES OC1=C(C(/C=C/C(O)=O)=CC=C1O)/C(C(O)=O)=C\C2=CC(O)=C(C=C2)O
Molecular Weight (g/mol) 358.3
Formula Weight 358.3
Percent Purity 96.81%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.