missing translation for 'onlineSavingsMsg'
Learn More

SMND-309, MedChemExpress

Product Code. 16498227
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498227 5 mg 5mg
16455518 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498227 Supplier Medchem Express Supplier No. HY13056

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SMND-309 is a metabolite of salvianolic acid B, and exhibits neuroprotective effects in cultured neurons and in permanent middle cerebral artery occlusion rats.

SMND-309 is a metabolite of salvianolic acid B, and exhibits neuroprotective effects in cultured neurons and in permanent middle cerebral artery occlusion rats.

Specifications

Chemical Name or Material SMND-309
CAS 1065559-56-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H14O8
Quantity 5 mg
Solubility Information DMSO : ≥ 3.7 mg/mL (10.33 mM)
SMILES OC1=C(C(/C=C/C(O)=O)=CC=C1O)/C(C(O)=O)=C\C2=CC(O)=C(C=C2)O
Molecular Weight (g/mol) 358.3
Formula Weight 358.3
Percent Purity 96.81%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.