missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMN Antibody (SMN1/1596), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44620-100ug
This item is not returnable.
View return policy
Description
SMN Monoclonal antibody specifically detects SMN in Human samples. It is validated for Immunohistochemistry
Specifications
| SMN | |
| Monoclonal | |
| 0.2 mg/ml | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry | |
| SMN1/1596 | |
| Unconjugated | |
| Gemin-1, Kugelberg-Welander disease), SMNC, survival motor neuron protein, survival of motor neuron 1, telomeric | |
| Recombinant full-length human SMN protein (Uniprot: Q16637). | |
| 100 μg | |
| Apoptosis, Neurodegeneration, Neuroscience, Transcription Factors and Regulators | |
| 6606 | |
| Store at 4⪚C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction