missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smilagenin, MedChemExpress
Description
Smilagenin (SMI) improves memory of aged rats by increasing the muscarinic receptor subtype 1 (M1)-receptor density. Smilagenin (SMI) attenuates Aβ(25-35)-induced neurodegenerationvia stimulating the gene expression of brain-derived neurotrophic factor, may represents a novel therapeutic strategy for AD.
Specifications
Specifications
| Chemical Name or Material | Smilagenin |
| CAS | 126-18-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C27H44O3 |
| Quantity | 50 mg |
| Solubility Information | Ethanol : ≥ 10 mg/mL (24.00 mM) ∣DMSO : 5 mg/mL (12.00 mM; Need ultrasonic) |
| SMILES | C[C@@H]1[C@@]2(CC[C@@H](C)CO2)O[C@]3([H])[C@@]1([H])[C@@]4([C@@]([C@@]5([H])[C@]([C@@]6([C@](C[C@@H](O)CC6)([H])CC5)C)([H])CC4)([H])C3)C |
| Molecular Weight (g/mol) | 416.64 |
| Formula Weight | 416.64 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?