missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMG5 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-47272PE
This item is not returnable.
View return policy
Description
SMG5 Polyclonal antibody specifically detects SMG5 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)
Specifications
| SMG5 | |
| Polyclonal | |
| Immunoprecipitation, Western Blot (Negative) | |
| EST1BFLJ12287, EST1-like protein B, Est1p-like protein B, ever shorter telomeres 1B, hSMG-5, KIAA1089FLJ34864, LPTS interacting protein, LPTS-interacting protein, LPTSRP1, LPTS-RP1EST1 telomerase component homolog B, protein SMG5, RP11-54H19.7, SMG-5, SMG-5 homolog, Smg-5 homolog, nonsense mediated mRNA decay factor (C. elegans) | |
| The immunogen for this product maps to a region between residue 400 and 450 of human Suppressor with Morphogenetic Effects on Genitalia 5 using the numbering given in entry NP_056142.2 (GeneID 23381). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 23381 | |
| Store at 4C in the dark. | |
| IgG |
| Immunoprecipitation | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction