missing translation for 'onlineSavingsMsg'
Learn More

SMG5 Antibody, Novus Biologicals™

Product Code. 18366812 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18366812 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18366812 Supplier Novus Biologicals Supplier No. NBP305264

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SMG5 Polyclonal specifically detects SMG5 in Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMG5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias EST1BFLJ12287, EST1-like protein B, Est1p-like protein B, ever shorter telomeres 1B, hSMG-5, KIAA1089FLJ34864, LPTS interacting protein, LPTS-interacting protein, LPTSRP1, LPTS-RP1EST1 telomerase component homolog B, protein SMG5, RP11-54H19.7, SMG-5, SMG-5 homolog, Smg-5 homolog, nonsense mediated mRNA decay factor (C. elegans)
Host Species Rabbit
Immunogen Recombinant protein of human SMG5.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 23381
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.