missing translation for 'onlineSavingsMsg'
Learn More

SMG5 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18433386 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433386 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433386 Supplier Novus Biologicals Supplier No. NBP147272FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SMG5 Polyclonal antibody specifically detects SMG5 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMG5
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EST1BFLJ12287, EST1-like protein B, Est1p-like protein B, ever shorter telomeres 1B, hSMG-5, KIAA1089FLJ34864, LPTS interacting protein, LPTS-interacting protein, LPTSRP1, LPTS-RP1EST1 telomerase component homolog B, protein SMG5, RP11-54H19.7, SMG-5, SMG-5 homolog, Smg-5 homolog, nonsense mediated mRNA decay factor (C. elegans)
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 400 and 450 of human Suppressor with Morphogenetic Effects on Genitalia 5 using the numbering given in entry NP_056142.2 (GeneID 23381).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 23381
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.