missing translation for 'onlineSavingsMsg'
Learn More

SMG5 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-47272DL594

Product Code. 18467486

  • £492.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SMG5 Polyclonal antibody specifically detects SMG5 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SMG5
Polyclonal
Western Blot
EST1BFLJ12287, EST1-like protein B, Est1p-like protein B, ever shorter telomeres 1B, hSMG-5, KIAA1089FLJ34864, LPTS interacting protein, LPTS-interacting protein, LPTSRP1, LPTS-RP1EST1 telomerase component homolog B, protein SMG5, RP11-54H19.7, SMG-5, SMG-5 homolog, Smg-5 homolog, nonsense mediated mRNA decay factor (C. elegans)
The immunogen for this product maps to a region between residue 400 and 450 of human Suppressor with Morphogenetic Effects on Genitalia 5 using the numbering given in entry NP_056142.2 (GeneID 23381).
0.1 mL
Cell Cycle and Replication
23381
Store at 4°C in the dark.
IgG
Western Blot
DyLight 594
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.