missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMG1 Antibody (SR1684), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22354-100ul
This item is not returnable.
View return policy
Description
SMG1 Monoclonal antibody specifically detects SMG1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SMG1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1684 | |
| Western Blot 1:500-1:2000 | |
| ATXserine/threonine-protein kinase SMG1, EC 2.7.11, EC 2.7.11.1, hSMG-1,61E3.4, KIAA0421phosphatidylinositol 3-kinase-related protein kinase, Lambda/iota protein kinase C-interacting protein, Lambda-interacting protein, LIPPI-3-kinase-related kinase SMG-1, phosphatidylinositol 3-kinase-related kinase, SMG-1, SMG1 homolog, phosphatidylinositol 3-kinase-related kinase (C. elegans) | |
| A synthesized peptide derived from human SMG1 (Uniprot #: Q96Q15) | |
| 100 μg | |
| Protein Kinase | |
| 23049 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction