missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies SMER28
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
- SMER28, a bromo-substituted quinazoline, was first identified as a small-molecule enhancer of rapamycin (SMER; Sarkar et al.) SMER28 induces autophagy by increasing autophagosome synthesis and enhancing the clearance of model autophagy substrates (Renna et al.)
Specifications
Specifications
| CAS | 307538-42-7 |
| Quantity | 10 mg |
| Molecular Formula | C11H10BrN3 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction