missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMEK1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566644
This item is not returnable.
View return policy
Description
Immunogen sequence: KLRFEQQRERQ DNPKLDSMRS ILRNHRYRRD ARTLEDEEEM WFNTDEDDME Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
Regulatory subunit of serine/threonine-protein phosphatase 4. May regulate the activity of PPP4C at centrosomal microtubule organizing centers. The PPP4C-PPP4R2-PPP4R3A PP4 complex specifically dephosphorylates H2AFX phosphorylated on 'Ser-140' (gamma-H2AFX) generated during DNA replication and required for DNA DSB repair.
Specifications
| SMEK1 | |
| Polyclonal | |
| Unconjugated | |
| PPP4R3A | |
| 1110034C04Rik; BC064465; FLFL1; KIAA2010; mKIAA2010; MSTP033; PP4R3; PP4R3A; Ppp4r3a; protein phosphatase 4 regulatory subunit 3A; protein phosphatase 4, regulatory subunit 3A; RGD1309059; serine/threonine-protein phosphatase 4 regulatory subunit 3A; SMEK homolog 1; SMEK homolog 1, suppressor of mek1; SMEK homolog 1, suppressor of mek1 (Dictyostelium); SMEK1; smk1; smk-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55671 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6IN85 | |
| PPP4R3A | |
| Recombinant Human SMEK1. Recombinant protein control fragment (Product #RP-105293). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction