missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smek1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98483B
This item is not returnable.
View return policy
Description
Smek1 Polyclonal specifically detects Smek1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| Smek1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| FLFL1, FLJ20707, KIAA2010MSTP033, PP4R3A, PPP4R3A, serine/threonine-protein phosphatase 4 regulatory subunit 3A, SMEK homolog 1, SMEK homolog 1, suppressor of mek1 (Dictyostelium), smk1, smk-1 | |
| Produced in rabbits immunized with E. coli-derived Human Smek1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Biotin | |
| PBS | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 55671 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction