missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SMCy 5.5

Product Code. 18768696
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18768696

Brand: Tocris Bioscience™ 7295/100U

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fluorogenic fluorescent probe for imaging lipid droplets; suitable for use with two-photon microscopy

SMCy 5.5 is a fluorogenic fluorescent probe for imaging lipid droplets that exhibits sharp excitation and emission bands and high brightness. SMCy 5.5 is suitable for use with two-photon microscopy. It is useful for deep tissue imaging and 3D imaging. Excitation/emission maxima for SMCy 5.5 in THF are 643/672 nm; Quantum yield = 0.75; Extinction coefficient = 209,000 M-1 cm-1. SMCy 5.5 displays high two-photon absoprtion (TPA) cross-section values (13330 GM and 10400 GM at 770 and 820 nm, respectively). The probe also has a click reactive handle.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6,8-Dicyclohexyl-4-((1 E,3 E,5 E)-5-(1,1-dimethyl-3-(pent-4-yn-1-yl)-1,3-dihydro-2 H-benzo[e]indol-2-ylidene)penta-1,3-dien-1-yl)-2,2-difluoro-2,8-dihydro-5 H-1λ3,2λ4-[1,3,2 ]dioxaborinino[6,5-d]pyrimidine-5,7(6 H)-dione
CAS 2417507-09-4
Quantity 100 μg
Target Fluorescent Lipids Probes and Cell Membrane Stains
Molecular Formula C41H46BF2N3O4
Purity 0.95
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.