missing translation for 'onlineSavingsMsg'
Learn More

SMCX Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18396392
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396392 100 μg 100µL
18314322 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18396392 Supplier Bio-Techne Supplier No. NBP318659100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SMCX Polyclonal antibody specifically detects SMCX in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMCX
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:500
Formulation Whole antiserum
Gene Accession No. P41229
Gene Alias DXS1272EMRXJ, EC 1.14.11, EC 1.14.11.-, Histone demethylase JARID1C, JARID1Clysine-specific demethylase 5C, jumonji, AT rich interactive domain 1C, Jumonji, AT rich interactive domain 1C (RBP2-like), Jumonji/ARID domain-containing protein 1C, lysine (K)-specific demethylase 5C, MRXSJ, Protein SmcX, Protein Xe169, Smcx homolog, X chromosome, SMCXselected cDNA on X, Smcy homolog, X-linked, Smcy homolog, X-linked (mouse), XE169JmjC domain-containing protein SMCX
Host Species Rabbit
Immunogen Polyclonal antibody raised in rabbit against mouse SMCX (jumonji, AT rich interactive domain 1C), using 3 KLH-conjugated synthetic peptides containing sequences from the inner part of the protein.
Purification Method Whole Antiserum
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8242
Target Species Human, Mouse
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Antisera
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.